Assay Details
Target Gene Details
Entrez Gene ID: | 244853 |
Gene Name: | neurexophilin and PC-esterase domain family, member 4 |
Gene Aliases: |
C130036J11, D930028F11Rik, Fam55d |
Location: |
Chr.9:48162001-48400025 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nxpe4 | AK021362.1 | |||
| AK048132.1 | BAC33253.1 | |||
| BC094249.1 | AAH94249.1 |

Genomic Map