Assay Details
Target Gene Details
Entrez Gene ID: | 215194 |
Gene Name: | KRI1 homolog (S. cerevisiae) |
Gene Aliases: |
- |
Location: |
Chr.9:21273457-21287969 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kri1 | NM_145416.3 | 14 | 1343 | NP_663391.2 |
| AK030447.1 | 1 | 1358 | ||
| BC021438.1 | 14 | 1333 | AAH21438.1 | |
| CO426749.1 | 6 | 575 | ||
| CX562209.1 | 4 | 370 |

Genomic Map