Assay Details
Target Gene Details
Entrez Gene ID: | 69797 |
Gene Name: | RIKEN cDNA 1600029I14 gene |
Gene Aliases: |
- |
Location: |
Chr.9:99470421-99474751 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1600029I14Rik | NR_028123.1 | 1 | 439 | |
| AK005560.1 | 1 | 419 | BAB24120.1 |
Target Gene Details
Entrez Gene ID: | 623534 |
Gene Name: | NME/NM23 family member 9 |
Gene Aliases: |
Txndc6 |
Location: |
Chr.9:99456243-99479899 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nme9 | NM_001165957.1 | 8 | 678 | NP_001159429.1 |

Genomic Map