Assay Details
Target Gene Details
Entrez Gene ID: | 72826 |
Gene Name: | family with sequence similarity 76, member B |
Gene Aliases: |
2810485I05Rik, C78303 |
Location: |
Chr.9:13827702-13854687 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam76b | NM_176836.3 | NP_789806.1 | ||
| AK052430.1 | ||||
| AK145026.1 | BAE26192.1 | |||
| BC043120.1 | AAH43120.1 | |||
| BC050870.1 | AAH50870.1 |
More Information
Set Membership: |
|

Genomic Map