Assay Details
Target Gene Details
Entrez Gene ID: | 382097 |
Gene Name: | predicted gene 1123 |
Gene Aliases: |
AV052824, CLSP |
Location: |
Chr.9:99006965-99035690 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm1123 | NM_001080776.1 | NP_001074245.1 | ||
| AB262662.1 | ||||
| BC114986.1 | AAI14987.1 | |||
| BC115956.1 |

Genomic Map