Assay Details
Target Gene Details
Entrez Gene ID: | 330962 |
Gene Name: | solute carrier family 51, beta subunit |
Gene Aliases: |
Ostb |
Location: |
Chr.9:65412753-65422773 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc51b | NM_178933.2 | NP_849264.1 | ||
| AK002463.1 | 1 | 109 | BAE20415.1 | |
| AK004188.1 | 1 | 161 | BAE20418.1 | |
| AY279396.1 | AAP23994.1 |
More Information
Set Membership: |
|

Genomic Map