Assay Details
Target Gene Details
Entrez Gene ID: | 70784 |
Gene Name: | RAS-like, family 12 |
Gene Aliases: |
4631404I11Rik |
Location: |
Chr.9:65398485-65415060 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rasl12 | NM_001162923.1 | NP_001156395.1 | ||
| AK048176.1 |
Target Gene Details
Entrez Gene ID: | 330962 |
Gene Name: | solute carrier family 51, beta subunit |
Gene Aliases: |
Ostb |
Location: |
Chr.9:65412753-65422773 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc51b | NM_178933.2 | 4 | 322 | NP_849264.1 |
| AK002463.1 | 4 | 453 | BAE20415.1 | |
| AK004188.1 | 4 | 505 | BAE20418.1 | |
| AY279396.1 | 4 | 322 | AAP23994.1 | |
| BC115606.1 | 3 | 301 | ||
| BC115607.1 | 3 | 301 |
More Information
Set Membership: |
|

Genomic Map