Assay Details
Target Gene Details
Entrez Gene ID: | 15481 |
Gene Name: | heat shock protein 8 |
Gene Aliases: |
2410008N15Rik, Hsc70, Hsc71, Hsc73, Hsp73, Hspa10 |
Location: |
Chr.9:40801273-40805199 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |

Genomic Map