Assay Details
Target Gene Details
Entrez Gene ID: | 331026 |
Gene Name: | GDP-mannose pyrophosphorylase B |
Gene Aliases: |
AI317178, E430010H19 |
Location: |
Chr.9:108049229-108051936 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gmppb | NM_177910.3 | 4 | 557 | NP_808578.1 |
| AK088295.1 | 4 | 539 | BAC40266.1 | |
| AK158470.1 | 4 | 557 | BAE34527.1 | |
| AK165816.1 | 4 | 559 | ||
| BC061207.1 | 5 | 431 | AAH61207.1 |
More Information
Set Membership: |
|

Genomic Map