Assay Details
Target Gene Details
Entrez Gene ID: | 235627 |
Gene Name: | neurobeachin-like 2 |
Gene Aliases: |
1110014F23Rik, BC042396, mKIAA0540 |
Location: |
Chr.9:110624789-110654161 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 52 - Exon 53 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nbeal2 | NM_183276.2 | NP_899099.2 | ||
| AK003694.2 | BAC25048.1 | |||
| AK129158.1 | BAC97968.2 | |||
| BC157956.1 | ||||
| BC172118.1 |

Genomic Map