Assay Details
Target Gene Details
Entrez Gene ID: | 66257 |
Gene Name: | nicolin 1 |
Gene Aliases: |
1500032A17Rik |
Location: |
Chr.9:108290457-108296496 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nicn1 | NM_025449.3 | NP_079725.1 | ||
| AJ299741.1 | CAC82517.1 | |||
| AK002789.1 | BAB22360.1 | |||
| AK013602.1 | BAB28922.1 | |||
| AK075732.1 | BAC35916.1 | |||
| AK146890.1 | BAE27508.1 | |||
| AK169135.1 | BAE40914.1 | |||
| BC024050.1 | AAH24050.1 |
More Information
Set Membership: |
|

Genomic Map