Assay Details
Target Gene Details
Entrez Gene ID: | 235587 |
Gene Name: | poly (ADP-ribose) polymerase family, member 3 |
Gene Aliases: |
A930002C11Rik, AW990611, Adprt3, Adprtl3, PARP-3, pADPRT-3 |
Location: |
Chr.9:106470353-106476944 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Parp3 | NM_145619.2 | NP_663594.2 | ||
| AF368233.1 | AAN62793.1 | |||
| AK044223.1 | BAC31826.1 | |||
| AK088125.1 | ||||
| AK145259.1 | BAE26330.1 | |||
| AK147868.1 | BAE28191.1 | |||
| AK165070.1 | ||||
| AK170541.1 | BAE41867.1 | |||
| BC014870.1 | AAH14870.1 | |||
| BC058754.1 | AAH58754.1 |
More Information
Set Membership: |
|

Genomic Map