Assay Details
Target Gene Details
Entrez Gene ID: | 245020 |
Gene Name: | solute carrier family 35, member G2 |
Gene Aliases: |
Gm519, Tmem22 |
Location: |
Chr.9:100552188-100571085 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc35g2 | NM_001101483.1 | NP_001094953.1 | ||
| BY726562.1 | ||||
| CB588129.1 |
More Information
Set Membership: |
|

Genomic Map