Assay Details
Target Gene Details
Entrez Gene ID: | 59092 |
Gene Name: | poly(rC) binding protein 4 |
Gene Aliases: |
1200003L19Rik, AlphaCP-4 |
Location: |
Chr.9:106453406-106464012 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcbp4 | NM_021567.5 | NP_067542.3 | ||
| AF176328.1 | AAG09239.1 | |||
| AK004580.1 | ||||
| AK005383.1 | BAB23990.1 | |||
| AK077632.1 | BAC36915.1 | |||
| AK082005.1 | ||||
| AK178757.1 | ||||
| AK179721.1 | ||||
| AK179793.1 | ||||
| AK179802.1 | ||||
| AK187694.1 | ||||
| AK188206.1 | ||||
| AK188263.1 | ||||
| AK189619.1 | ||||
| BC010694.1 | AAH10694.1 |
More Information
Set Membership: |
|

Genomic Map