Assay Details
Target Gene Details
Entrez Gene ID: | 66904 |
Gene Name: | propionyl Coenzyme A carboxylase, beta polypeptide |
Gene Aliases: |
1300012P06Rik, AI314687, R74805 |
Location: |
Chr.9:100982032-101034940 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pccb | NM_025835.2 | NP_080111.1 | ||
| AF327060.1 | AAK28525.1 | |||
| AK004976.1 | BAB23712.1 | |||
| AK050366.1 | BAC34211.1 | |||
| AK134437.1 | BAE22142.1 | |||
| BC026531.1 | ||||
| BC037082.1 | AAH37082.1 | |||
| BC055946.1 | AAH55946.1 |
More Information
Set Membership: |
|

Genomic Map