Assay Details
Target Gene Details
Entrez Gene ID: | 56289 |
Gene Name: | Ras association (RalGDS/AF-6) domain family member 1 |
Gene Aliases: |
123F2, AA536941, AU044980, NORE2A, RDA32, REH3P21, Rassf1A, Rassf1B, Rassf1C |
Location: |
Chr.9:107551555-107562267 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rassf1 | NM_001243748.1 | NP_001230677.1 | ||
| NM_019713.4 | NP_062687.1 | |||
| AF132851.1 | AAD30061.1 | |||
| AK146736.1 | BAE27398.1 | |||
| BC002173.1 | AAH02173.1 | |||
| CD564987.1 | ||||
| CX224234.1 | ||||
| EU314810.1 |
More Information
Set Membership: |
|

Genomic Map