Assay Details
Target Gene Details
Entrez Gene ID: | 100568458 |
Gene Name: | predicted gene, 38431 |
Gene Aliases: |
Raver1-fdx1l |
Location: |
Chr.9:21067514-21092008 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm38431 | NR_038081.2 | |||
| AK004919.1 | BAB23670.1 | |||
| AY275472.1 | AAP33691.1 |
Target Gene Details
Entrez Gene ID: | 71766 |
Gene Name: | ribonucleoprotein, PTB-binding 1 |
Gene Aliases: |
1300006N24Rik, AA050359, mKIAA1978 |
Location: |
Chr.9:21074158-21091988 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Raver1 | NM_027911.3 | NP_082187.1 | ||
| AK051444.1 | ||||
| AK079191.1 | ||||
| AK084478.1 | BAC39194.1 | |||
| AK138596.1 | ||||
| AK168436.1 | ||||
| BC058271.1 | ||||
| BC108392.1 | AAI08393.1 |
More Information
Set Membership: |
|

Genomic Map