Assay Details
Target Gene Details
Entrez Gene ID: | 331026 |
Gene Name: | GDP-mannose pyrophosphorylase B |
Gene Aliases: |
AI317178, E430010H19 |
Location: |
Chr.9:108049229-108051936 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gmppb | NM_177910.3 | NP_808578.1 | ||
| AK158470.1 | BAE34527.1 | |||
| AK165816.1 |
Target Gene Details
Entrez Gene ID: | 84585 |
Gene Name: | ring finger protein 123 |
Gene Aliases: |
BC003945, Kpc1 |
Location: |
Chr.9:108051672-108079773 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 38 - Exon 39 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnf123 | NM_032543.2 | NP_115932.1 | ||
| AK137599.1 | ||||
| AY744153.1 | AAU93471.1 | |||
| BC057082.1 | AAH57082.1 |
More Information
Set Membership: |
|

Genomic Map