Assay Details
Target Gene Details
Entrez Gene ID: | 102634431 |
Gene Name: | predicted gene, 32014 |
Gene Aliases: |
- |
Location: |
Chr.9:8644078-8740988 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm32014 | NR_110513.1 | |||
| AK054106.1 |
Target Gene Details
Entrez Gene ID: | 22068 |
Gene Name: | transient receptor potential cation channel, subfamily C, member 6 |
Gene Aliases: |
AV025995, LLHWJM002, LLHWJM003, LLHWJM004, TRP-6, Trrp6, mtrp6 |
Location: |
Chr.9:8543932-8680752 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trpc6 | NM_001282086.1 | 7 | 2109 | NP_001269015.1 |
| NM_013838.2 | 8 | 2343 | NP_038866.2 | |
| AF057748.1 | 8 | 1912 | AAC64394.1 | |
| AK133864.1 | 7 | 2009 | BAE21896.1 | |
| AK137447.1 | 7 | 1571 | ||
| BC141131.1 | 8 | 2214 | ||
| U49069.1 | 8 | 2343 | AAC06146.1 |
More Information
Set Membership: |
|

Genomic Map