Assay Details
Target Gene Details
Entrez Gene ID: | 21838 |
Gene Name: | thymus cell antigen 1, theta |
Gene Aliases: |
CD90, T25, Thy-1, Thy-1.2, Thy1.1, Thy1.2 |
Location: |
Chr.9:44043384-44048579 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Thy1 | NM_009382.3 | NP_033408.1 | ||
| AK080366.1 | 1 | 803 | ||
| AY445633.1 | AAR17087.1 | |||
| BC054436.1 | AAH54436.1 | |||
| CX123560.1 | ||||
| M12380.1 | AAO25750.1 |
More Information
Set Membership: |
|

Genomic Map