Assay Details
Target Gene Details
Entrez Gene ID: | 77862 |
Gene Name: | thymocyte nuclear protein 1 |
Gene Aliases: |
D730042P09Rik, HSPC144, MDS012, MY105, Thy28 |
Location: |
Chr.9:26999659-27007334 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Thyn1 | NM_144543.2 | 3 | 281 | NP_653126.1 |
| AB080370.1 | 2 | 205 | BAC21100.1 | |
| AK021335.1 | 3 | 249 | ||
| AK077639.1 | 3 | 251 | BAC36919.1 | |
| BC016594.1 | 3 | 235 | AAH16594.1 | |
| CB236103.1 | 2 | 474 |

Genomic Map