Assay Details
Target Gene Details
Entrez Gene ID: | 77862 |
Gene Name: | thymocyte nuclear protein 1 |
Gene Aliases: |
D730042P09Rik, HSPC144, MDS012, MY105, Thy28 |
Location: |
Chr.9:26999659-27007334 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Thyn1 | NM_144543.2 | 7 | 710 | NP_653126.1 |
| AB080370.1 | 6 | 634 | BAC21100.1 | |
| AK077639.1 | BAC36919.1 | |||
| BC016594.1 | 7 | 664 | AAH16594.1 |
Target Gene Details
Entrez Gene ID: | 69091 |
Gene Name: | vacuolar protein sorting 26 homolog B (yeast) |
Gene Aliases: |
1810012I05Rik, 2310075A12Rik, AI848392 |
Location: |
Chr.9:27004502-27030094 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vps26b | NM_178027.4 | 6 | 4500 | NP_821170.1 |
| AK040818.1 | 1 | 890 | ||
| AK041580.1 | 1 | 538 | ||
| AK163860.1 | 1 | 2203 |
More Information
Set Membership: |
|

Genomic Map