Assay Details
Target Gene Details
Entrez Gene ID: | 18746 |
Gene Name: | pyruvate kinase, muscle |
Gene Aliases: |
AA414905, AL024370, AL024424, Pk-2, Pk-3, Pk3, Pkm2 |
Location: |
Chr.9:59656368-59679375 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pkm | NM_001253883.1 | NP_001240812.1 | ||
| NM_011099.3 | NP_035229.2 | |||
| AK083076.1 | ||||
| AK135397.1 | BAE22519.1 | |||
| AK151724.1 | BAE30642.1 | |||
| AK153483.1 | BAE32031.1 | |||
| AK154695.1 | ||||
| AK155110.1 | BAE33055.1 | |||
| AK155655.1 | BAE33370.1 | |||
| AK168943.1 | BAE40751.1 | |||
| AK169872.1 | ||||
| AK170892.1 | BAE42098.1 | |||
| AK171023.1 | BAE42192.1 | |||
| AK171033.1 | BAE42199.1 | |||
| AK171106.1 | ||||
| AK200951.1 | ||||
| AK210659.1 | ||||
| BC016619.1 | AAH16619.1 | |||
| BC094663.1 | AAH94663.1 | |||
| CB846337.1 | ||||
| D38379.1 | BAA07457.1 |
More Information
Set Membership: |
|

Genomic Map