Assay Details
Target Gene Details
Entrez Gene ID: | 70984 |
Gene Name: | RIKEN cDNA 4931406C07 gene |
Gene Aliases: |
- |
Location: |
Chr.9:15283337-15306448 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4931406C07Rik | NM_001199484.1 | 1 | 96 | NP_001186413.1 |
| AK133192.1 | 1 | 97 |
Target Gene Details
Entrez Gene ID: | 75316 |
Gene Name: | TATA box binding protein (Tbp)-associated factor, RNA polymerase I, D |
Gene Aliases: |
2810003M17Rik, 4930553M18Rik, 5930426I11Rik, Josd3, TAF(I)41, TAFI41 |
Location: |
Chr.9:15306202-15316997 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Taf1d | NM_027261.3 | NP_081537.1 | ||
| NM_029248.2 | NP_083524.2 | |||
| NR_028401.1 | ||||
| AK012662.1 | BAB28391.1 | |||
| AK014920.1 | BAB29621.1 | |||
| AK016109.1 | BAB30118.1 | |||
| AK050165.1 | BAC34104.1 | |||
| AK158208.1 | 1 | 95 | ||
| AK161656.1 | ||||
| AK166329.1 | BAE38709.1 | |||
| AK166633.1 | ||||
| BU850945.1 |
More Information
Set Membership: |
|

Genomic Map