Assay Details
Target Gene Details
Entrez Gene ID: | 21345 |
Gene Name: | transgelin |
Gene Aliases: |
Sm22, Sm22a, Ws310 |
Location: |
Chr.9:45929628-45936058 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tagln | NM_011526.5 | 2 | 322 | NP_035656.1 |
| AK002880.1 | 2 | 161 | BAB22427.1 | |
| AK077235.1 | 2 | 163 | BAC36700.1 | |
| BC003795.1 | 2 | 151 | AAH03795.1 | |
| L41154.1 | 2 | 161 | AAA79166.1 | |
| U36588.1 | 2 | 161 | AAC52418.1 |
More Information
Set Membership: |
|

Genomic Map