Assay Details
Target Gene Details
Entrez Gene ID: | 55934 |
Gene Name: | retinitis pigmentosa 9 (human) |
Gene Aliases: |
PAP-1, Rp9h |
Location: |
Chr.9:22448311-22468356 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rp9 | NM_018739.2 | 1 | 117 | NP_061209.1 |
| AK146136.1 | 1 | 99 | BAE26927.1 | |
| AK168391.1 | 1 | 110 | BAE40317.1 | |
| BC115549.1 | 1 | 77 | ||
| BC115550.1 | 1 | 77 | ||
| BY239981.1 | 1 | 117 | ||
| D78255.1 | 1 | 96 | BAA11319.1 |

Genomic Map