Assay Details
Target Gene Details
Entrez Gene ID: | 60507 |
Gene Name: | queuine tRNA-ribosyltransferase 1 |
Gene Aliases: |
2610028E17Rik, Tgt |
Location: |
Chr.9:21411572-21420279 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Qtrt1 | NM_021888.2 | NP_068688.2 | ||
| AB034632.1 | BAA93550.1 | |||
| AK011586.1 | BAB27717.1 | |||
| BC044811.1 | AAH44811.1 | |||
| BY291101.1 |
More Information
Set Membership: |
|

Genomic Map