Assay Details
Target Gene Details
Entrez Gene ID: | 26611 |
Gene Name: | reticulocalbin 2 |
Gene Aliases: |
AA408742, Tcbp49 |
Location: |
Chr.9:56041845-56059083 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rcn2 | NM_011992.2 | 3 | 524 | NP_036122.2 |
| AF049125.1 | 3 | 510 | AAC05132.1 | |
| AK076153.1 | 3 | 351 | ||
| AK077486.1 | 3 | 500 | BAC36825.1 | |
| AK078106.1 | 3 | 492 | ||
| AK169765.1 | 3 | 498 | BAE41353.1 | |
| BB638743.1 | 3 | 521 | ||
| BC132320.1 | 3 | 489 | ||
| BC145668.1 | 3 | 489 |
More Information
Set Membership: |
|

Genomic Map