Assay Details
Target Gene Details
Entrez Gene ID: | 67891 |
Gene Name: | ribosomal protein L4 |
Gene Aliases: |
2010004J23Rik |
Location: |
Chr.9:64173387-64178562 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpl4 | NM_024212.4 | NP_077174.1 | ||
| AK002361.1 | ||||
| AK007625.1 | ||||
| AK008098.1 | BAB25458.1 | |||
| AK011068.1 | BAB27375.1 | |||
| AK012432.1 | BAB28234.2 | |||
| AK050639.1 | ||||
| AK088054.1 | ||||
| AK088279.1 | BAC40254.1 | |||
| AK135186.1 | ||||
| AK151950.1 | ||||
| AK165962.1 | BAE38489.1 | |||
| AK166748.1 | BAE38990.1 | |||
| AK166957.1 | BAE39140.1 | |||
| AK167054.1 | BAE39218.1 | |||
| BC003459.1 | AAH03459.1 | |||
| BC094036.1 | AAH94036.1 | |||
| BC132321.1 | ||||
| BC132323.1 |
More Information
Set Membership: |
|

Genomic Map