Assay Details
Target Gene Details
Entrez Gene ID: | 80385 |
Gene Name: | tumor suppressor candidate 2 |
Gene Aliases: |
1190001E22Rik, AA407686, Fus1, Lgcc, PAP, Pdap2 |
Location: |
Chr.9:107563255-107566108 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tusc2 | NM_019742.4 | NP_062716.1 | ||
| AF123387.1 | AAD42286.1 | |||
| AK004430.1 | BAB23300.1 | |||
| AK082684.1 | BAC38571.1 | |||
| BC048477.1 | AAH48477.1 |

Genomic Map