Assay Details
Target Gene Details
Entrez Gene ID: | 629557 |
Gene Name: | glyceraldehyde-3-phosphate dehydrogenase pseudogene |
Gene Aliases: |
EG629557 |
Location: |
Chr.9:52002061-52048705 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm6981 | NR_023357.1 | |||
| AK132984.1 |
Target Gene Details
Entrez Gene ID: | 19684 |
Gene Name: | radixin |
Gene Aliases: |
AA516625 |
Location: |
Chr.9:52047150-52088738 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rdx | NM_001104616.1 | NP_001098086.1 | ||
| DV662898.1 |
More Information
Set Membership: |
|

Genomic Map