Assay Details
Target Gene Details
Entrez Gene ID: | 67941 |
Gene Name: | ribosomal protein S27-like |
Gene Aliases: |
1810034D23Rik |
Location: |
Chr.9:66946076-66949520 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rps27l | NM_026467.3 | NP_080743.1 | ||
| AK007689.1 | BAB25192.1 | |||
| AK011262.1 | BAB27503.1 | |||
| AK151083.1 | BAE30096.1 | |||
| BC058115.1 | AAH58115.1 |
More Information
Set Membership: |
|

Genomic Map