Assay Details
Target Gene Details
Entrez Gene ID: | 16785 |
Gene Name: | ribosomal protein SA |
Gene Aliases: |
67kDa, 67lr, AL022858, Lamr, Lamr1, Lamrl1, MLR, P40, P40-3, P40-8 |
Location: |
Chr.9:120127766-120132369 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
More Information
Set Membership: |
|

Genomic Map