Assay Details
Target Gene Details
Entrez Gene ID: | 23999 |
Gene Name: | twinfilin, actin-binding protein, homolog 2 (Drosophila) |
Gene Aliases: |
A6-related, AU014993, Ptk9l, Ptk9r |
Location: |
Chr.9:106203108-106215387 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Twf2 | NM_011876.3 | NP_036006.1 | ||
| AK002699.1 | 1 | 42 | BAB22293.1 | |
| AK003997.1 | ||||
| AK084026.1 | BAC39101.1 | |||
| AK087422.1 | 1 | 42 | BAC39867.1 | |
| AK155204.1 | BAE33117.1 | |||
| AK157886.1 | BAE34246.1 | |||
| AK166900.1 | BAE39103.1 | |||
| AK170441.1 | BAE41799.1 | |||
| AY267189.1 | AAP31405.1 | |||
| BC003338.1 | AAH03338.1 | |||
| Y17808.1 | CAB38083.1 |
More Information
Set Membership: |
|

Genomic Map