Assay Details
Target Gene Details
Entrez Gene ID: | 22771 |
Gene Name: | zinc finger protein of the cerebellum 1 |
Gene Aliases: |
ZIC, ZNF201 |
Location: |
Chr.9:91358058-91365826 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zic1 | NM_009573.3 | NP_033599.2 | ||
| AK136977.1 | BAE23194.1 | |||
| AK141883.1 | BAE24869.1 | |||
| AK159769.1 | BAE35357.1 | |||
| BC050889.1 | ||||
| BC060247.1 | AAH60247.1 | |||
| BC063247.1 | AAH63247.1 | |||
| D32167.1 | BAA06878.1 |
Target Gene Details
Entrez Gene ID: | 22774 |
Gene Name: | zinc finger protein of the cerebellum 4 |
Gene Aliases: |
- |
Location: |
Chr.9:91361777-91389349 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zic4 |
More Information
Set Membership: |
|

Genomic Map