Assay Details
Target Gene Details
Entrez Gene ID: | 12696 |
Gene Name: | cold inducible RNA binding protein |
Gene Aliases: |
Cirp, R74941 |
Location: |
Chr.10:80165990-80172691 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cirbp | NM_007705.2 | NP_031731.1 | ||
| AK014655.1 | BAB29491.1 | |||
| AK132584.1 | BAE21245.1 | |||
| BC075699.1 | AAH75699.1 | |||
| D78135.1 | BAA11213.1 |

Genomic Map