Assay Details
Target Gene Details
Entrez Gene ID: | 12215 |
Gene Name: | basigin |
Gene Aliases: |
AI115436, AI325119, CD147, EMMPRIN, HT-7 |
Location: |
Chr.10:79704358-79711979 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bsg | NM_001077184.1 | NP_001070652.1 | ||
| NM_009768.2 | NP_033898.1 | |||
| AK002332.1 | BAB22018.1 | |||
| AK011364.1 | ||||
| AK147225.1 | ||||
| AK207023.1 | ||||
| AK208329.1 | ||||
| AK209680.1 | ||||
| AK214501.1 | ||||
| AK215038.1 | ||||
| AK216955.1 | ||||
| AY089967.1 | AAM09957.1 | |||
| BC010270.1 | AAH10270.1 | |||
| D00611.1 | BAA00486.1 | |||
| S63813.1 | AAB27567.2 | |||
| Y16256.1 | CAA76140.1 | |||
| Y16257.1 | CAA76141.1 | |||
| Y16258.1 | CAA76142.1 |
More Information
Set Membership: |
|

Genomic Map