Assay Details
Target Gene Details
Entrez Gene ID: | 77048 |
Gene Name: | centrosomal protein 83 |
Gene Aliases: |
2600001G24Rik, 4921537D05Rik, 5730513H21Rik, Ccdc41 |
Location: |
Chr.10:94688495-94790339 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cep83 | NM_029852.2 | NP_084128.2 | ||
| AK015007.1 | BAB29669.2 | |||
| AK135868.1 | BAE22706.1 | |||
| AK151291.1 | BAE30276.1 | |||
| AK152465.1 | BAE31242.1 | |||
| AK152801.1 | BAE31506.1 | |||
| AK153515.1 | BAE32058.1 | |||
| BC043468.1 | AAH43468.1 | |||
| BC070464.1 | AAH70464.1 |

Genomic Map