Assay Details
Target Gene Details
Entrez Gene ID: | 77048 |
Gene Name: | centrosomal protein 83 |
Gene Aliases: |
2600001G24Rik, 4921537D05Rik, 5730513H21Rik, Ccdc41 |
Location: |
Chr.10:94688495-94790339 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cep83 | NM_029852.2 | 3 | 595 | NP_084128.2 |
| AK011135.1 | 3 | 667 | ||
| AK015007.1 | 3 | 732 | BAB29669.2 | |
| AK141627.1 | 3 | 716 | BAE24774.1 | |
| BC043468.1 | 2 | 209 | AAH43468.1 | |
| BC070464.1 | 3 | 595 | AAH70464.1 |

Genomic Map