Assay Details
Target Gene Details
Entrez Gene ID: | 76306 |
Gene Name: | solute carrier family 18, subfamily B, member 1 |
Gene Aliases: |
1110021L09Rik |
Location: |
Chr.10:23796654-23827969 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc18b1 | NM_183116.2 | 3 | 386 | NP_898939.2 |
| AK183134.1 | 3 | 296 | ||
| CK625631.1 | 3 | 316 | ||
| CX201878.1 |
More Information
Set Membership: |
|

Genomic Map