Assay Details
Target Gene Details
Entrez Gene ID: | 76306 |
Gene Name: | solute carrier family 18, subfamily B, member 1 |
Gene Aliases: |
1110021L09Rik |
Location: |
Chr.10:23796654-23827969 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc18b1 | NM_183116.2 | NP_898939.2 | ||
| AK184257.1 | ||||
| AK184817.1 | ||||
| AK217807.1 | ||||
| BM949736.1 | ||||
| CX201878.1 |
More Information
Set Membership: |
|

Genomic Map