Assay Details
Target Gene Details
Entrez Gene ID: | 73218 |
Gene Name: | signal peptide peptidase like 2B |
Gene Aliases: |
3110056O03Rik, AW550292, PSL1 |
Location: |
Chr.10:80855248-80868708 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sppl2b | NM_175195.3 | 6 | 755 | NP_780404.2 |
| AK046804.1 | 6 | 774 | BAC32878.1 | |
| AK148109.1 | 6 | 722 | BAE28349.1 | |
| AK157906.1 | 6 | 700 | ||
| AK165324.1 | 6 | 880 | ||
| AK166364.1 | 6 | 706 | BAE38734.1 | |
| AK170375.1 | 6 | 721 | BAE41755.1 | |
| BC052094.1 | 4 | 429 | AAH52094.1 |
More Information
Set Membership: |
|

Genomic Map