Assay Details
Target Gene Details
Entrez Gene ID: | 73218 |
Gene Name: | signal peptide peptidase like 2B |
Gene Aliases: |
3110056O03Rik, AW550292, PSL1 |
Location: |
Chr.10:80855248-80868708 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sppl2b | NM_175195.3 | NP_780404.2 | ||
| AK046804.1 | BAC32878.1 | |||
| AK148109.1 | BAE28349.1 | |||
| AK157906.1 | ||||
| AK165324.1 | ||||
| AK166364.1 | BAE38734.1 | |||
| AK170375.1 | BAE41755.1 | |||
| AK187931.1 | ||||
| BC052094.1 | AAH52094.1 |
More Information
Set Membership: |
|

Genomic Map