Assay Details
Target Gene Details
Entrez Gene ID: | 52666 |
Gene Name: | Rho guanine nucleotide exchange factor (GEF) 25 |
Gene Aliases: |
2410008H17Rik, D10Ertd610e, GEFT |
Location: |
Chr.10:127182521-127190088 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgef25 | NM_001166413.1 | NP_001159885.1 | ||
| NM_028027.3 | NP_082303.2 | |||
| AF487515.1 | AAO49464.1 | |||
| AK010452.1 | BAB26951.1 | |||
| AK158684.1 | BAE34610.1 | |||
| AK210287.1 | ||||
| CF724487.1 |
More Information
Set Membership: |
|

Genomic Map