Assay Details
Target Gene Details
Entrez Gene ID: | 11630 |
Gene Name: | absent in melanoma 1 |
Gene Aliases: |
AI462491, AI463325 |
Location: |
Chr.10:43950307-44148924 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aim1 | NM_172393.2 | 9 | 4424 | NP_765981.2 |
| AF397913.1 | 3 | 131 | ||
| AK041082.1 | 8 | 1040 | ||
| AK088102.1 | 8 | 1096 | BAC40145.1 | |
| BC035551.1 | 8 | 1576 | AAH35551.1 |

Genomic Map