Assay Details
Target Gene Details
Entrez Gene ID: | 208677 |
Gene Name: | cAMP responsive element binding protein 3-like 3 |
Gene Aliases: |
BC010786, CREB-H, D10Bur1e |
Location: |
Chr.10:81084324-81098872 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Creb3l3 | NM_145365.3 | NP_663340.1 | ||
| AF392874.1 | AAM73673.1 | |||
| AK050186.1 | BAC34115.1 | |||
| AK077258.1 | BAC36714.1 | |||
| BC010786.1 | AAH10786.1 | |||
| BC028820.1 | AAH28820.1 |

Genomic Map