Assay Details
Target Gene Details
Entrez Gene ID: | 12534 |
Gene Name: | cyclin-dependent kinase 1 |
Gene Aliases: |
Cdc2, Cdc2a, p34<CDC2> |
Location: |
Chr.10:69336635-69352912 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdk1 | NM_007659.3 | NP_031685.2 | ||
| AK030231.1 | BAC26856.1 | |||
| AK051859.1 | ||||
| AK135516.1 | BAE22561.1 | |||
| AK168054.1 | BAE40034.1 | |||
| BC005614.1 | AAH05614.1 | |||
| BC024396.1 | AAH24396.1 | |||
| M38724.1 | AAA37408.1 | |||
| U58633.1 | AAB09465.1 | |||
| X16461.1 | CAA34481.1 |
More Information
Set Membership: |
|

Genomic Map