Assay Details
Target Gene Details
Entrez Gene ID: | 544696 |
Gene Name: | TBC1 domain family, member 32 |
Gene Aliases: |
Bromi, D630037F22Rik, b2b2284Clo |
Location: |
Chr.10:56014293-56229456 on Build GRCm38 |
Assay Gene Location: | Within Exon 32 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbc1d32 | NM_001033385.3 | 32 | 3748 | NP_001028557.2 |
| AK035346.1 | 14 | 1528 |

Genomic Map