Assay Details
Target Gene Details
Entrez Gene ID: | 66337 |
Gene Name: | family with sequence similarity 229, member B |
Gene Aliases: |
1700025K23Rik |
Location: |
Chr.10:39118804-39133943 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam229b | NM_183254.1 | 3 | 253 | NP_899077.1 |
| AK006353.1 | 3 | 253 | BAC25140.1 | |
| AK006431.1 | 3 | 231 | ||
| BC147171.1 | 2 | 182 | ||
| BC147172.1 | 2 | 182 | ||
| BC172004.1 | 2 | 179 |

Genomic Map