Assay Details
Target Gene Details
Entrez Gene ID: | 14219 |
Gene Name: | connective tissue growth factor |
Gene Aliases: |
Ccn2, Fisp12, Hcs24, fisp-12 |
Location: |
Chr.10:24595442-24598683 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ctgf | NM_010217.2 | NP_034347.2 | ||
| BC006783.1 | AAH06783.1 | |||
| BQ917383.1 | ||||
| M70642.1 | AAA37628.1 | |||
| M80263.1 | AAA73135.1 |
More Information
Set Membership: |
|

Genomic Map